Ashcroft IXLDP Ultra-Low Differential Pressure Transmitters
  • FEATURES
  • Certified 0.25% and 0.50% accuracy
  • High over pressure protection
  • NEMA 4X metal construction
  • Six output signals
  • Easy installation
  • FM approved for hazardous locations (optional)
Designed for the measurement and control of very low pressure and flow in industrial and process plant environments. The transmitter features s rugged NEMA4X enclosure, built-in electrical terminal box isolated from the electronics and threaded process connections.
$946.00 - $1,305.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Accuracy Max Pressure (PSI) Resolution
Availability Pricing
Ashcroft IXLDP5IWC Low Diff Pressure Transmitter, 5.0" WC, 4-20mA, 0.25% EW-68439-31 Mfr # IX-3-F02-42-ST-5IW
COMPARE
±0.25% full-scale
$1,305.00USD / Each
Ashcroft IXLDP5IWC Low Diff Pressure Transmitter, 5.0" WC, 4-20mA, 0.5% EW-68439-43 Mfr # IX-5-F02-42-ST-5IW
COMPARE
±0.5% full-scale
$946.00USD / Each