Ashcroft® Model 1008 Stainless Steel Pressure Gauges
Ideal for pumps and compressors, hydraulic and pneumatics applications, specialized OEM equipment, or food and beverage
  • Corrosion resistant stainless steel case
  • Dry gauges are field fillable
  • PowerFlex™ movement provides superior resistance to shock, vibration, and pulsation
  • True Zero™ reduces reading errors by using “zero box” instead of conventional dial pins
  • MSL helium leak tested to 1X10-6 scc/s
  • Ventable fill plug
Available in 2.5" (63 mm) and 3.5" (100 mm) dial sizes, Ashcroft® 1008S pressure gauges are field liquid fillable and field convertible for panel mounting. ASME Grade B, 3-2-3% accuracy is standard.
ITEM(S) ON SALE
$51.28 - $54.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Ashcroft 1008 Stainless Steel Pressure Gauge, 2.5" Dial, 1/4" NPT (M) Lower Connection, 30" Hg to 15 psi EW-68022-42 Mfr # 631008S 02L VAC/0/15#
COMPARE
Stainless Steel Pressure Gauge, 2.5" Dial, 1/4" NPT (M) Lower Connection, 30" Hg to 15 psi
$54.00USD / Each
$67.50USD / Each
Ashcroft 1008 Stainless Steel Pressure Gauge, 2.5" Dial, 1/4" NPT (M) Lower Connection, 0 to 60 psi EW-68022-45 Mfr # 631008S 02L 0/60#
COMPARE
Stainless Steel Pressure Gauge, 2.5" Dial, 1/4" NPT (M) Lower Connection, 0 to 60 psi
$52.96USD / Each
$66.20USD / Each
Ashcroft 1008 Stainless Steel Pressure Gauge, 2.5" Dial, 1/4" NPT (M) Lower Connection, 0 to 100 psi EW-68022-46 Mfr # 631008S 02L 0/100#
COMPARE
Stainless Steel Pressure Gauge, 2.5" Dial, 1/4" NPT (M) Lower Connection, 0 to 100 psi
$51.28USD / Each
$64.10USD / Each