Bürkle Disposable HDPE Sampling Ladles
Easily pour from either side with dual-spout design
  • Ergonomically designed handle for ease of use
  • Manufactured in cleanroom environment and individually packaged
  • Constructed of durable HDPE for chemically aggressive applications
Disposable HDPE sampling ladles are suitable for use with foodstuff. Cup has a capacity of 200 mL and features embossed graduations in increments of 10 mL up to 200 mL. The ergonomic handle and spout on both sides of the ladle allow for right- or left-handed operation. Individually packaged; available nonsterile or sterilized via gamma irradiation.
$199.50 - $346.00USD / Pkg of 20
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description Length (in) Sample Volume
Availability Pricing
Burkle 5393-6640 Disposable sampling scoop, HDPE, 500 mm handle; 200 mL EW-06043-51 Mfr # 5393-6640
COMPARE
Disposable Sampling Scoop, HDPE, 200 mL, 50 cm Length; 20/pk 19.7 200 mL
$199.50USD / Pkg of 20
Burkle 5393-6650 Disposable sampling scoop, HDPE, 1000 mm handle; 200 mL EW-06043-53 Mfr # 5393-6650
COMPARE
Disposable Sampling Scoop, HDPE, 200 mL, 100 cm Length; 20/pk 39.4 200 mL
$244.00USD / Pkg of 20
Burkle 5393-6651 Disposable sampling scoop, HDPE, 1000 mm handle, sterile; 200 mL EW-06043-54 Mfr # 5393-6651
COMPARE
Disposable Sampling Scoop, HDPE, 200 mL, 100 cm Length, Sterile; 20/pk 39.4 200 mL
$346.00USD / Pkg of 20