Bacharach Informant® 2 Refrigerant and Combustible Leak Detectors
Don’t wait for results—fastest and most reliable meter
  • Swap sensor tips in seconds with no need to adjust sensitivity
  • Sensors are the industry’s most responsive, reliable, and accurate
  • Hydrophobic filter prevents moisture contamination
  • Sequential flashing probe tip, dual LED displays and loud tick rate indicate relative concentration levels
  • Detects all halogens and scores of hydrocarbon gases
  • Get into hard-to-reach areas with tapered refigerant sensor tip— just 0.2" diameter
Its unique combination of state-of-the-art sensor technology, advanced software design, and microprocessor-controlled circuitry offers you unmatched precision coupled with tremendous speed. It is unquestionably the most sensitive, most reliable, most accurate leak detector on the market today. And the fastest. Swap sensor tips within seconds and never worry about adjusting sensitivity levels — it's taken care of automatically, even where high levels of background vapors or refrigerants are present.
The leak detector features an audible indicator with mute option, coupled with sequential LEDs to monitor increasing concentrations of refrigerant or combustible gases. Dual bar graph displays offer visual confirmation no matter what position the instrument is in. And a flashing light at the tip of the probe helps to visually define your sampling location.
$897.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Gas Measured Probe
Availability Pricing
Bacharach 0019-8038 Contractor Refrigerant/Combustile Gas Leak Kit EW-10110-45 Mfr # H0019-8038
COMPARE
Methane 20" (50.8 cm) fully extended; 3" (7.62 cm) if wrapped around instrument
$897.00USD / Each