BactiZapper Micro-Sterilizer
  • Sterilizes platinum loops and needles safely and conveniently
  • Prevents infectious spatter and cross contamination
The BactiZapper utilizes infrared heat to incinerate organic material deep within the ceramic funnel. Sterilization takes only 5 to 7 seconds at optimum sterilizing temperature of 1500°F. Unit can be used in anaerobic chambers
$764.00 - $784.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC)
Availability Pricing
Benchmark Scientific BactiZapper Infrared Sterilizer, 120 VAC EW-01850-22 Mfr # B1000
COMPARE
120
$764.00USD / Each
Benchmark Scientific BactiZapper Infrared Sterilizer, 230 VAC EW-01850-24 Mfr # B1000-E
COMPARE
230
$784.00USD / Each