BrandTech® Discharge Tube Kit for Dispensette® S Dispensers
Perfect for filling racks of tubes
  • Not suitable for HF
  • PTFE wetted material
  • Features a recirculation valve
  • Approximately 800 mm L coiled tubing with safety handle
  • Textured handle provides a secure grip
Use the flexible discharge tube kit with Dispensette® S dispensers for serial dispensing. A built-in valve help rid away with bubbles.
$273.00 - $291.50USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Compatible With
Availability Pricing
BrandTech 708132 Flexible Discharge Tube Kit for Dispensette® S Dispensers, PTFE, 1, 2, 5, or 10 mL EW-07915-98 Mfr # 708132
COMPARE
Dispensette® S dispenser with nominal volume of 1, 2, 5, or 10 mL
$273.00USD / Each
BrandTech 708134 Flexible Discharge Tube Kit for Dispensette® S Dispensers, PTFE, 25, 50, or 100 mL EW-07915-99 Mfr # 708134
COMPARE
Dispensette® S dispenser with nominal volume of 25, 50, or 100 mL
$291.50USD / Each