BrandTech® Pipette Tips for Air Displacement Pipettes

Graduated for quick volume check (excluding 5 & 10 mL sizes)
  • Manufactured without lubricants
  • Cadmium-free pigments
  • Autoclavable at 121 °C
Designed to fit BRAND pipettes and other pipettes using non-proprietary tips, these tips are polypropylene free from oleamide and diHEMDA - additives which can interfere with biological tests. Pipette tips are produced using the most advanced cleanroom techniques to ensure consistently high quality. Bulk-packed in reclosable bags.
$58.70 - $114.50USD / Pkg of 2000

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Volume (mL) Package Of
Availability Pricing
BrandTech 732004 Pipette Tips, 0.5 to 20 uL, Bulk; 2000/PK EW-24553-31 Mfr # 732004
COMPARE
0.02 Bulk; 2000/Pk
$99.10USD / Pkg of 2000
BrandTech 702595 Pipette Tips, 0.5 to 5 mL, Bulk; 200/PK EW-24553-34 Mfr # 702595
COMPARE
5 Bulk; 200/Bag
$58.70USD / Bag of 200
BrandTech 702603 Pipette Tips, 1 to 10 mL, Bulk; 200/PK EW-24553-35 Mfr # 702603
COMPARE
10 Bulk; 200/Bag
$114.50USD / Pkg of 200
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat