BrandTech® Threaded Bottle Adapters for Dispensers and Digital Burettes
Adapt dispenser to bottles with various thread sizes for your application needs
  • Easily transition from your dispenser to your bottle
$13.90 - $53.50USD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Connections Material
Availability Pricing
BrandTech 704399 Threaded Bottle Adapter, ETFE, GL45 Thread x GL38 Bottle Thread EW-07902-08 Mfr # 704399
COMPARE
GL45 Thread x GL38 Bottle Thread ETFE
$47.00USD / Each
BrandTech 704398 Threaded Bottle Adapter, ETFE, GL45 Thread x GL32 Bottle Thread EW-07910-07 Mfr # 704398
COMPARE
GL45 Thread x GL32 Bottle Thread ETFE
$47.00USD / Each
BrandTech 704325 Threaded Bottle Adapter, PP, GL32 Thread x GL25 Bottle Thread EW-07910-78 Mfr # 704325
COMPARE
GL32 Thread x GL25 Bottle Thread Polypropylene
$13.90USD / Each
BrandTech 704328 Threaded Bottle Adapter, PP, GL32 Thread x GL28 Bottle Thread EW-07910-79 Mfr # 704328
COMPARE
GL32 Thread x GL28 Bottle Thread Polypropylene
$13.90USD / Each
BrandTech 704345 Threaded Bottle Adapter, PP, GL32 Thread x GL45 Bottle Thread EW-07910-87 Mfr # 704345
COMPARE
GL32 Thread x GL45 Bottle Thread Polypropylene
$13.90USD / Each
BrandTech 704396 Threaded Bottle Adapter, PP, GL45 Thread x GL32 Bottle Thread EW-07910-95 Mfr # 704396
COMPARE
GL45 Thread x GL32 Bottle Thread Polypropylene
$13.90USD / Each
BrandTech 704397 Threaded Bottle Adapter, PP, GL45 Thread x GL38 Bottle Thread EW-07910-96 Mfr # 704397
COMPARE
GL45 Thread x GL38 Bottle Thread Polypropylene
$13.90USD / Each
BrandTech 704429 Threaded Bottle Adapter, PP, GL32 Thread x NS29/32 (Ground Joint) EW-07910-99 Mfr # 704429
COMPARE
GL32 Thread x NS29/32 (Ground Joint) Polypropylene
$53.50USD / Each
Showing 1- 8 of 8