Burkle TeleScoop Telescoping Open Container Sampler, Beakers and Bottle Holder
Versatile, secure, and easy to use
  • Attach different beakers and bottles to suit any sampling application
  • User-friendly snap-in joint is practical and efficient in the field or on the floor
This modular sampling system allows you to build a sampler according to your application. Select a telescoping aluminum rod (not included) with the length you need, then choose one or more beakers and/or a bottle holder to attach to the rod. Angled and pendulum beakers are available in plastic or metal, and the bottle holder fits plastic, glass, or metal bottles.
Angled beakers maintain position while sampling. The polypropylene beakers feature an adjustable angle up to 90°, and the 304 SS beaker has a fixed angle with a lip—perfect for scraping caked solids, sludge, and mud.
Pendulum beakers automatically swings into a horizontal position, preventing spills. This type of beaker is ideal for deep, narrow shafts and other difficult-to-reach areas.
Bottle holder uses a universal quick-tightening strap to hold bottles up to 88 mm in diameter securely during sampling. Base supports bottom of bottle and provides an adjustable angle.
$87.10 - $344.50USD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Material Volume (mL) Description
Availability Pricing
Burkle 5354-0050 Beaker Sampler, PP and Adjustable Metal Holder,  600 mL EW-06041-99 Mfr # 5354-0050
COMPARE
Polypropylene (PP) - Beaker 600 Beaker Sampler, PP and Adjustable Metal Holder, 600 mL
$93.30USD / Each
Burkle 5354-0100 Beaker Sampler, PP and Adjustable Metal Holder, 1000 mL EW-06042-00 Mfr # 5354-0100
COMPARE
Polypropylene (PP) - Beaker 1000 Beaker Sampler, PP and Adjustable Metal Holder, 1000 mL
$103.00USD / Each
Burkle 5354-0200 Beaker Sampler, PP and Adjustable Metal Holder, 2000 mL EW-06042-01 Mfr # 5354-0200
COMPARE
Polypropylene (PP) - Beaker 2000 Beaker Sampler, PP and Adjustable Metal Holder, 2000 mL
$113.50USD / Each
Burkle 5354-2000 Beaker Sampler for Telescopic Rod, SS; 1000 mL EW-06042-03 Mfr # 5354-2000
COMPARE
304 SS 1000 Angled Beaker for TeleScoop, Fixed, 304 SS; 1000 mL
$315.50USD / Each
Burkle 5354-3000 Pendulum Beaker, SS; 1000 mL EW-06042-04 Mfr # 5354-3000
COMPARE
304 SS 1000 Pendulum Beaker for TeleScoop, 304 SS; 1000 mL
$344.50USD / Each
Burkle 5354-4100 Bottle Holder, PP/PA
Always in Stock
EW-06042-05 Mfr # 5354-4100
COMPARE
Polypropylene (PP), Polyamide (PA) 750 Bottle Holder for TeleScoop, Adjustable Up to 88 mm Dia., PP/PA; Max 750 mL
$131.00USD / Each
Burkle 5354-5050 Pendulum Beaker, PP; 600 mL EW-06042-06 Mfr # 5354-5050
COMPARE
Polypropylene (PP) 600 Pendulum Beaker for TeleScoop, PP; 600 mL
$87.10USD / Each
Burkle 5354-5100 Pendulum Beaker, PP; 1000 mL EW-06042-07 Mfr # 5354-5100
COMPARE
Polypropylene (PP) 1000 Pendulum Beaker for TeleScoop, PP; 1000 mL
$91.50USD / Each
Showing 1- 8 of 8