CAPP ecopipette Single-Channel Variable-Volume Pipettors
Go green with this environmentally-friendly and economically-priced line of pipettors!
  • Recycled or recyclable materials used for packaging
  • Durable construction and long warranty period decreases waste
From the moment you recieve your new CAPP ecopipette you will notice the difference. The packaging is made entirely of recycled or recyclable materials such as post-consumer paper or lightweight cardboard for reduced environmental impact.
The pipettor itself is designed and manufactured to stand the test of time. The aluminum tip cone is resistant to breakage and will not deform like the lower quality plastics used in other pipettors. This combined with the double o-ring design provides a better fit with a broader range of pipette tips. The three-year warranty ensures longterm performance and reliabilty which minimizing discarded pipettors going into landfills.
And while you are going green, you can save some "green" by taking advantage of the very attractive pricing on these quality name-brand adjustable volume pipettors. Color-coded control knobs make it easy to identify which volume range you have on the pipettor body. The control knob can be inserted in either direction to allow right or left handed operation. Since the volume display is on the control knob, your fingers do not obscure the display from sight. Volume setting is easy and requires half the number of turns compared to other pipettors. The volume locking device prevents unwanted volume changes.
Ergonomic features include very low pressure required to depress the plunger for aspiration, dispensing, and blowout; large tip ejector pad; and a comfortable index finger loop. Ecopipette bodies are fully autoclavable without disassembly.
$458.00 - $477.50USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Code Max Volume (µL) Min Volume (µL)
Availability Pricing
Capp Ecopipette Adjustable-Volume Pipettor, 20-200 µL; Each EW-25201-14 Mfr # C200-1
COMPARE
Yellow 200 20
$458.00USD / Each
Capp Ecopipette Adjustable-Volume Pipettor, 100-1000 µL; Each EW-25201-16 Mfr # C1000-1
COMPARE
Blue 1000 100
$477.50USD / Each