Cole-Parmer® Autoclavable Biohazard Bags
Heavy-gauge, 2.25 mil thick polypropylene (PP) bags offer excellent puncture resistance
  • Withstand temperatures up to 136°C (277°F)
  • Prominently marked with the words “Hazardous Waste” and the “Biohazard” symbol
  • White marking area for quick and easy labeling
These high-temperature autoclavable bags are extremely pliable yet offer exceptional strength and durability. Seamless bottoms ensure leak-free autoclaving. Opaque "biohazard" red color along with the words "Hazardous Waste" and the "biohazard" symbol conforms to OSHA standard (29CFR1910.1030). An oversized white marking area makes it easy to label each bag with a permanent marker.
$37.00 - $110.00USD / Pkg of 100
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (Gal) Length (in) Width (in)
Availability Pricing
COMPARE
3 26 12.2
$110.00USD / Case of 200
COMPARE
0.43 11 8.5
$37.00USD / Pkg of 100