Cole-Parmer® Bulb Pipette Fillers
Thicker rubber walls offer greater pipetting control
  • Manufactured for the safe and reliable aspiration and delivery of liquids
  • Ideal for use with serological pipettes up to 50mL
Natural rubber pipette filler is ideal for serological pipettes with 50-mL capacity. Special pinch and release valve system with a chemical-resistant stainless steel ball bearing allows for easy one-hand operation. Available in a variety of color choices.
$11.50USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Color Code Wetted Matterial Description
Availability Pricing
COMPARE
Black Rubber and Stainless Steel Bulb Pipette Filler, Up to 50 mL, Black
$11.50USD / Each
COMPARE
Green Rubber and Stainless Steel Bulb Pipette Filler, Up to 50 mL, Green
$11.50USD / Each