Cole-Parmer® Cellulose Acetate Filters
Ideal for filtering biological solutions
  • Exhibit low protein binding
Filters are Triton-free and have a bidirectional membrane support. Membrane type and pore size are printed on each filter for easy identification. All Cole-Parmer syringe filters are 100% integrity tested and manufactured in accordance with ISO 9002 standards. They are certified nonpyrogenic and noncytotoxic.
$147.20 - $231.00USD / Box of 50
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Membrane Diameter (mm) Pore Size (µm) Sterile
Availability Pricing
COMPARE
26 0.45 Yes
$188.80USD / Box of 50
COMPARE
26 0.8 Yes
$189.85USD / Box of 50
COMPARE
28 0.2 Yes
$195.85USD / Box of 50
COMPARE
25 0.45 No
$147.20USD / Box of 100
COMPARE
25 0.2 No
$147.20USD / Box of 100
COMPARE
26 0.8 No
$231.00USD / Box of 100