Cole-Parmer® Citric Acid, ACS Grade

Reagents and solutions to meet your lab and research needs
  • Quality and consistency your lab can count on
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$54.10 - $210.00USD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
CAS Number Weight (g) Description
Availability Pricing
COMPARE
77-92-9 1000 Citric Acid, ACS Grade, Anhydrous, 1000 g
$137.00USD / Each
COMPARE
77-92-9 100 Citric Acid, ACS Grade, Anhydrous, 100 g
$54.10USD / Each
COMPARE
77-92-9 500 Citric Acid, ACS Grade, Anhydrous, 500 g
$80.80USD / Each
COMPARE
5949-29-1 1000 Citric Acid, ACS Grade, Monohydrate, 1000 g
$210.00USD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat