Cole-Parmer® Cryo-Sentry Liquid Nitrogen Low-Level Alarms
A visual and an audible alarms alert you of low liquid nitrogen levels
  • Automatically checks the liquid level every 30 minutes
  • Connect an external alarm device to the relay output
  • Operates on AA batteries or AC power
Protect your valuable biological samples with the low-level alarm for your cryogenic storage dewars. The low-level alarm checks the liquid level every 30 minutes conserving battery power. Easily check the level without opening the vessel by pressing the LEVEL CHECK button.
$1,375.00 - $1,505.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Compatible With Description
Availability Pricing
COMPARE
Dewar 03773-04 Cryo-Sentry Low-Level Alarm for Dewar 03773-04
$1,375.00USD / Each
COMPARE
Dewar 03773-05 Cryo-Sentry Low-Level Alarm for Dewar 03773-05
$1,505.00USD / Each
COMPARE
Dewars 03773-06 and -07 Cryo-Sentry Low-Level Alarm for Dewars 03773-06 and -07
$1,505.00USD / Each
COMPARE
Dewars 03773-08 and -09 Cryo-Sentry Low-Level Alarm for Dewars 03773-08 and -09
$1,505.00USD / Each