Cole-Parmer® Disposable 2-Way, 3-Port Stopcocks with Luer Fittings, Polycarbonate
Restrict your flow to only two directions with the 2-way, 3-port stopcocks
  • Consistent inside diameter minimizes turbulence and bubble entrapment
  • Handle points to "off" port
  • Use with both liquids and gases
  • DEHP free
  • Polycarbonate (PC) housing and polyethylene rotor
Stopcocks provide an easy on/off control of your fluid. Two-way, three-port stopcocks feature two female Luer fittings and a male Luer fitting with or without a lock. Luer fittings on stopcocks ensure a quick, leak-free connection. Simply turn the handle to a port you want closed; the other two ports remain open. The handle can only be turned to the "off" position at the male Luer port or female side Luer port. Stopcocks are made of polyethylene rotor and transparent polycarbonate body, making it easy to see the flow.
$13.00 - $100.50USD / Pkg of 10
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Connection Type Luer Connection Type (Port 3) Quantity
Availability Pricing
COMPARE
Female Luer x Female Luer x Male Locking Luer Male Locking Luer 10
$13.80USD / Pkg of 10
COMPARE
Female Luer x Female Luer x Male Slip Luer Male Slip Luer 10
$13.00USD / Pkg of 10
COMPARE
Female Luer x Female Luer x Male Locking Luer Male Locking Luer 100
$100.50USD / Pkg of 100
COMPARE
Female Luer x Female Luer x Male Slip Luer Male Slip Luer 100
$94.20USD / Pkg of 100