Cole-Parmer® Economy Pre-Cleaned Wide-Mouth Bottle, Amber Glass
Saves valuable time—odor-free and tasteless finish will not affect stored products
  • Pre-cleaned by vacuum and ionization
  • Individually sealed for long term storage with no contamination
The vacuum and ionization process removes contaminants such as dirt, dust, carton lint, oil, aerosols, and fine glass particles from these bottles. Ideal for chemical and resin sampling. Bottles come with green thermoset, PTFE-lined caps.
$97.50 - $246.50USD / Case of 24
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Cap / Stopper Size Material
Availability Pricing
COMPARE
30 28-400 Glass
$97.50USD / Case of 24
COMPARE
60 33-400 Glass
$125.00USD / Case of 24
COMPARE
120 38-400 Glass
$150.00USD / Case of 24
COMPARE
250 45-400 Glass
$187.25USD / Case of 24
COMPARE
500 53-400 Glass
$124.00USD / Case of 12
COMPARE
1000 53-400 Glass
$246.50USD / Case of 12