Cole-Parmer® General Purpose Freestanding Dual Temperature Refrigerator/Freezer Combinations
Versatile cooling and freezing for general purpose applications
  • Adjustable refrigerator (1°C to 10°C) and freezer (-15°C to -25°C) settings
  • Secure and efficient design with a keyed door lock, recessed handles, and a magnetic door gasket for a reliable seal
  • Uses natural hydrocarbon refrigerants for a sustainable choice
Cole-Parmer® General Purpose Freestanding Dual Temperature Refrigerator/Freezer Combinations provide an ideal solution for everyday storage needs where advanced temperature precision and monitoring are not required. With an adjustable mechanical thermostat, these units offer flexible temperature ranges for both refrigeration and freezing. The freezer section includes both manual and auto defrost options to suit your maintenance preferences. Thoughtful design features such as recessed door handles, a keyed lock, and a magnetic gasket enhance usability and security. Environmentally conscious hydrocarbon refrigerants ensure compliance with sustainability goals, while a robust two-year parts and labor warranty delivers lasting peace of mind.
ITEM(S) ON SALE
$1,396.00 - $2,720.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Freezer Capacity (cu ft) Power (VAC) Refrigerator Capacity (cu ft)
Availability Pricing
COMPARE
1 120 3
$1,396.00USD / Each
$1,745.00USD / Each
COMPARE
5 120 15
$2,720.00USD / Each
$3,400.00USD / Each