Cole-Parmer® Graduated Polypropylene Beakers with Handle
Molded in handle provides easy transport and pouring of contents
  • Molded in graduations will not wear off
  • Accurate to ±3% to 4%
  • Beaker can withstands temperatures from 5 to 248°F (-15 to 150°C)
Polypropylene (PP) beakers with handles and pour spouts are ideal for general use in the lab.
$6.50 - $50.40USD / Pkg of 5
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Description
Availability Pricing
COMPARE
250 50 Plastic Beaker with Handle, Translucent PP, 250 mL, 5/pk
$6.50USD / Pkg of 5
COMPARE
500 50 Plastic Beaker with Handle, Translucent PP, 500 mL, 5/pk
$8.30USD / Pkg of 5
COMPARE
1000 50 Plastic Beaker with Handle, Translucent PP, 1000 mL, 5/pk
$13.70USD / Pkg of 5
COMPARE
2000 100 Plastic Beaker with Handle, Translucent PP, 2000 mL, 4/pk
$16.80USD / Pkg of 4
COMPARE
3000 50 Plastic Beaker with Handle, Translucent PP, 3000 mL, 5/pk
$38.70USD / Pkg of 5
COMPARE
5000 100 Plastic Beaker with Handle, Translucent PP, 5000 mL, 5/pk
$50.40USD / Pkg of 5