Cole-Parmer® Grain Spears
Reliably sample various powders, granules, and grains
  • Suitable for sampling from all layers of the container, providing an accurate representation of the contents
  • Constructed of lightweight aluminum for ease of use and durability
These grain samplers provide highly representative sampling of powders, granules, and grains. The spiral position of the pockets means the bottom pocket opens and fills first, giving a more representative sample. The lightweight aluminum construction means even the long samplers are easy to operate and manage. The tip is fixed permanently so it cannot be lost. The sampler is manufactured so that it can be stripped and cleaned within seconds. Sampler handle sold separately.
$398.00 - $413.50USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Core Diameter (in) Length (in) Max Sampling Depth (in)
Availability Pricing
COMPARE
1 3/8 58 29/64 42.91
$398.00USD / Each
COMPARE
1 3/8 78 5/32 57.09
$413.50USD / Each