Cole-Parmer® HDPE In-Line Filter, Polypropylene
Effectively removes visible particles, impurities, and contaminants
  • Suitable for a broad range of industrial and commercial applications
  • Built with a robust polypropylene housing for long-lasting performance
  • Equipped with hose barb fittings to fit various tubing sizes
These HDPE in-line filters are engineered to effectively remove visible particles, impurities, and contaminants from fluid systems. Suitable for a wide range of industries, these filters provide reliable performance across diverse applications.
Each filter features a durable polypropylene housing with hose barb fittings on both ends, ensuring compatibility with various tubing sizes. Available in 45–90 micron and 90–130 micron options, they offer versatile filtration to meet your specific needs.
$13.90 - $14.50USD / Pkg of 10
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connections Filter Size Membrane Material
Availability Pricing
COMPARE
1/8" ID Hose Barb 45-90 Micron HDPE
$13.90USD / Pkg of 10
COMPARE
1/8" ID Hose Barb 90-130 Micron HDPE
$13.90USD / Pkg of 10
COMPARE
3/16" ID Hose Barb 45-90 Micron HDPE
$13.90USD / Pkg of 10
COMPARE
3/16" ID Hose Barb 90-130 Micron HDPE
$14.20USD / Pkg of 10
COMPARE
1/4" ID Hose Barb 45-90 Micron HDPE
$14.20USD / Pkg of 10
COMPARE
1/4" ID Hose Barb 90-130 Micron HDPE
$14.20USD / Pkg of 10
COMPARE
3/8" ID Hose Barb 45-90 Micron HDPE
$14.50USD / Pkg of 10
COMPARE
3/8" ID Hose Barb 90-130 Micron HDPE
$14.50USD / Pkg of 10
Showing 1- 8 of 8