Cole-Parmer® High-Efficiency Fume Hoods
Large 29" high sash height allows ample room for loading, unloading, and use of equipment
  • VaraFlow baffle system maintains uniform airflow
Ducted fume hoods feature an ergonomic counter-balanced 3/16" tempered safety glass sash lift. Hoods are nonmetallic, composite fiberglass dual-wall construction and have an angled picture-frame style opening. The vapor-proof fluorescent lighting is wired to a preinstalled junction box with switch. Airflow monitor is also preinstalled and wired for immediate use. Included epoxy work surface features dished groove for spill capture and user safety. A 2-service, grounded, 20 amp outlet is also preinstalled for use with mixers, stirrers, or other laboratory equipment. Customize your hood with plumbing and gas fixtures; contact an Application Specialist for more information. Standard and acid-storage base cabinets are available under “Accessories”; 72" hoods require two 36" cabinets.
Note:All Hoods need to be hard-wired and require a blower for operation.
$24,345.00 - $24,355.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Exterior Height (in) Exterior Width (in) Power (VAC)
Availability Pricing
COMPARE
48 48 115
$24,345.00USD / Each
COMPARE
48 72 115
$24,355.00USD / Each