Cole-Parmer® Mortar and Pestle Sets, Agate
Ideal for laboratories that require contamination-free results!
  • Physical dimensions have a stated tolerance of ±3mm
  • Autoclavable
Sold as a set, these mortar and pestle sets provide you with optimal results and easy cleaning with highly polished grinding surface. Constructed with naturally hard agate stone - 6.5 hardness on Mohs scale.
$148.00 - $1,835.00USD / Set of 1
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Mortars Capacity (mL) Mortars Top OD (mm) Pestles Length (mm)
Availability Pricing
COMPARE
5 38 38
$148.00USD / Set of 1
COMPARE
10 50 65
$157.00USD / Set of 1
COMPARE
25 76 76
$269.75USD / Set of 1
COMPARE
50 89 89
$413.50USD / Set of 1
COMPARE
75 102 102
$482.00USD / Set of 1
COMPARE
250 152 152
$1,835.00USD / Set of 1