Cole-Parmer® MP-250 Series Digital Melting Point Apparatus
Rotating viewing head ensures each user can analyze samples clearly
  • View up to three samples simultaneously for greater efficiency
  • Height-adjustable arm with viewing magnifier folds away for easy storage
  • Integral light and 8x magnifying lens enhance sample observation
  • Three audible beeps indicate when temperature is stable and ready for sample
  • Adjustable object lens for sharp focus
The MP-250D series digital melting point apparatus allows you to easily view three capillary tube samples simultaneously through the 8x magnifying lens. The adjustable extension arm can be set to different viewing heights making it ideal for multiple users. A rotating viewing head and safety eye piece eliminate glare on the lens and protect you against hot zone.
A 13-key membrane keypad offers easy-to-use menus. The conveniently located push button controls allow temperature recording without looking away from the sample. A digital microprocessor with ±0.1°C resolution provides fast warm-up and accurate temperature control.
Units can be calibrated by on site or by the manufacturer/service organization. A calibration kit is required if you choose to calibrate on site.
$1,590.00 - $3,770.00USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Heat-Up Time Max Temperature (° C) Description
Availability Pricing
COMPARE
1° or 10°C per minute (selectable) 400 Standard Digital Melting Point Apparatus, 1°C Temperature Resolution; 100-240 VAC
$1,590.00USD / Each
COMPARE
0.2 to 10⁰C per minute 400 Programmable Digital Melting Point Apparatus; 100-240 VAC, 50/60 Hz
$2,325.00USD / Each
COMPARE
1° or 10°C per minute (selectable) 400 Standard Digital Melting Point Apparatus; 100-240 VAC, 50/60 Hz
$1,900.00USD / Each
COMPARE
0.2 to 10⁰C per minute 400 Programmable Digital Melting Point Apparatuses, Beginning/Ending Recording; 100-240 VAC
$2,325.00USD / Each
COMPARE
0.2 to 10⁰C per minute 400 Programmable Digital Melting Point Apparatus, Beginning/Ending Recording with IQ/OQ Documentation; 100-240 VAC
$3,770.00USD / Each