Cole-Parmer® MP-400 Series Digital Melting Point Apparatus
Digitally measure three melting points at once
  • Be more productive—test three samples simultaneously
  • Maximum temperature of 400°C with 0.1° resolution
  • Variable ramp rate between 0.5 and 10°C in 0.1°C increments after initial warm-up
  • Easily view the melt via magnifying lens with built-in illumination
  • Save up to seven temperature readings per capillary
  • Two-stage head adjustment offers user comfort
Select, measure, and display the temperature digitally, ensuring accuracy and avoiding the need for a glass thermometer. Test three samples simultaneously, viewing the melt via a magnifying lens with built-in illumination. Set a plateau temperature for initial warm-up—unattended warm-up time frees up the user until the plateau temperature is reached. Heating rate continues slowly between 0.5 and 10°C after the warm-up, slowly reaching the melt temperature for greater accuracy.
Save and recall up to seven temperatures per capillary. Three methods can be stored and repeated. Use the 5-pin mini DIN socket to connect to a printer (order separately). Adjust the viewing head two ways for better viewing and user comfort. The head can be moved forwards/backwards as well as tilted up to 126°. A convenient capillary storage compartment comes with a glass capillary cutter. Full access to the block allows for easy cleaning.
$2,805.00 - $3,000.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° C) Power (VAC) Temperature Resolution
Availability Pricing
COMPARE
400 115 / 230 0.1°C
$3,000.00USD / Each
COMPARE
400 115 / 230 0.1°C
$2,805.00USD / Each