Cole-Parmer® Nonsterile Syringe Filters, PVDF

Resistant to many solvents and show low protein binding—ideal for general filtration of biological samples
  • Broad chemical compatibility and low UV absorbing extractable
  • Solvent-resistant filter housing with nominal extractable
  • Leak-free Luer-Lok® and Luer-slip connections
  • Color-coded filters for easy organization
Prepare reliable aqueous, organic, and biomolecular samples with membrane filtration inside a polypropylene (PP) housing. Each housing comes with a color-coded retainer ring seal to prevent leaks and sample loss, and the color helps to visually show you the membrane type inside the housing.

Syringe filters have Luer-Lok and Luer-slip connections for easy, quick and tight connections to tubing, syringes, or needles. Filter packs are available in a resealable storage container to prevent contamination during multiple opening and closings.
$134.15 - $1,141.35USD / Pkg of 100

16 variations of this product are available.

See All Product Options Below
16 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (mm) Pore Size (µm) Description
Availability Pricing
COMPARE
0.22 Syringe Filter, PVDF, 4 mm dia., 0.22 µm; 100/pk
$135.45USD / Pkg of 100
COMPARE
0.22 Syringe Filter, PVDF, 4 mm dia., 0.22 µm; 1000/pk
$1,069.95USD / Pkg of 1000
COMPARE
0.45 Syringe Filter, PVDF, 4 mm dia., 0.45 µm; 100/pk
$138.60USD / Pkg of 100
COMPARE
0.45 Syringe Filter, PVDF, 4 mm dia., 0.45 µm; 1000/pk
$1,141.35USD / Pkg of 1000
COMPARE
0.22 Syringe Filter, PVDF, 13 mm dia., 0.22 µm; 100/pk
$134.15USD / Pkg of 100
COMPARE
0.22 Syringe Filter, PVDF, 13 mm dia., 0.22 µm; 1000/pk
$1,010.10USD / Pkg of 1000
COMPARE
0.45 Syringe Filter, PVDF, 13 mm dia., 0.45 µm; 100/pk
$143.35USD / Pkg of 100
COMPARE
0.45 Syringe Filter, PVDF, 13 mm dia., 0.45 µm; 1000/pk
$1,004.35USD / Pkg of 1000
COMPARE
0.22 Syringe Filter, PVDF, 25 mm dia., 0.22 µm; 100/pk
$136.50USD / Pkg of 100
COMPARE
0.22 Syringe Filter, PVDF, 25 mm dia., 0.22 µm; 500/pk
$607.95USD / Pkg of 500
COMPARE
0.45 Syringe Filter, PVDF, 25 mm dia., 0.45 µm; 100/pk
$141.00USD / Pkg of 100
COMPARE
0.45 Syringe Filter, PVDF, 25 mm dia., 0.45 µm; 500/pk
$631.05USD / Pkg of 500
COMPARE
0.22 Syringe Filter, PVDF, 30 mm dia., 0.22 µm; 100/pk
$157.50USD / Pkg of 100
COMPARE
0.22 Syringe Filter, PVDF, 30 mm dia., 0.22 µm; 500/pk
$622.65USD / Pkg of 500
COMPARE
0.45 Syringe Filter, PVDF, 30 mm dia., 0.45 µm; 100/pk
$139.15USD / Pkg of 100
COMPARE
0.45 Syringe Filter, PVDF, 30 mm dia., 0.45 µm; 500/pk
$612.70USD / Pkg of 500
Showing 1- 16 of 16

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat