Cole-Parmer® Polyethersulfone (PES) Filter Membranes
Remove particulate without adversely affecting your final product
  • Use membrane filtration for samples holding suspended solids
  • Highly efficient filtration of particles according to size
  • Complete sample filtration with the least sample loss and energy use
  • Filtration for HPLC sample preparation and mobile phase
  • Uniform manufacturing for repeatable results
$102.90 - $197.70USD / Pkg of 100
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (mm) Pore Size (µm)
Availability Pricing
COMPARE
25 0.22
$102.90USD / Pkg of 100
COMPARE
47 0.22
$187.45USD / Pkg of 100
COMPARE
47 0.45
$197.70USD / Pkg of 100