Cole-Parmer® Powder Lances, HDPE
Quickly and easily take a cross sectional sample within the vessel
  • Manufactured in Class 100,000 cleanroom and individually packaged
  • Ergonomically designed handle makes it easy to use, even when doing a large amount of sampling
  • Constructed of chemically inert high-density polyethylene (HDPE)
These powder lance samplers are designed to take a cross sectional sample at an exact position within the vessel. The samplers are manufactured, assembled, and packed in a Class 100,000 Cleanroom to ensure cleanliness and individually bagged. Choose from sterile or non-sterile samplers. Sterilized powder lances are sterilized via gamma irradiation (25-45 kGy).
$194.75 - $295.75USD / Pkg of 20
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (cm) Sterile Volume (mL)
Availability Pricing
COMPARE
95 No 200
$295.75USD / Pkg of 20
COMPARE
40 No 75
$194.75USD / Pkg of 20
COMPARE
40 Yes 75
$269.00USD / Pkg of 20
COMPARE
60 No 125
$241.25USD / Pkg of 20
COMPARE
60 Yes 125
$277.25USD / Pkg of 20