Cole-Parmer® Pre-Cleaned Narrow-Mouth Bottles, Clear Glass
Save time – no need to prewash bottles
  • Pre-cleaned by vacuum and ionization
The vacuum and ionization process removes contaminants such as dirt, dust, carton lint, oil, aerosols, and fine glass particles from bottles. Ideal for chemical and resin sampling. Individually sealed for long term storage with no contamination.
$94.50 - $147.75USD / Case of 48
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Cap / Stopper Size Material
Availability Pricing
COMPARE
30 20 mm Glass
$147.75USD / Case of 48
COMPARE
125 22 mm Glass
$103.00USD / Case of 24
COMPARE
500 28 mm Glass
$94.50USD / Case of 12
COMPARE
1000 33 mm Glass
$122.00USD / Case of 12