Cole-Parmer® Sleeve Sample Chamber and Handle
Reliably sample large volumes at great depths
  • Designed to be extremely strong and robust to allow samples to be taken from depths up to 6 meters
  • Constructed of durable 316L stainless steel
The sleeve sampler is ideal for sampling large volumes of powder or granules at a depth of up to 6 meters. The heavy duty 316L stainless steel handle provides enhanced strength and durability. Simply push the sleeve sampler into the powder or granules to the required depth and pull up on the handle to open the sleeve and take a sample.
Please Note: Sleeve sampler handle and chamber sold separately.
$329.00 - $397.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Material Description
Availability Pricing
COMPARE
7.48 316L Stainless Steel Sample Chamber, Stainless Steel, 100 mL
$397.00USD / Each
COMPARE
42.13 316L Stainless Steel Sleeve Sampler Handle, Stainless Steel, 1000 mm Length
$329.00USD / Each