Cole-Parmer® Slot Samplers, Metal
Reliably sample various powders, granules, and grains
  • Suitable for sampling from all layers of the container, providing an accurate representation of the contents
  • Choose from stainless steel or aluminum construction
These slot samplers provide highly representative sampling of powders, granules, and grains. They are suitable for sampling from all layers of the container, providing an accurate representation of the contents. These samplers come in different lengths, with different number of openings, and made of different material options for all different sampling methods.
The lightweight aluminum construction means even the long samplers are easy to operate and manage. The 316L stainless steel construction is durable and ideal when chemical resistance is required.
$229.75 - $1,385.00USD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (cm) Material Volume (mL)
Availability Pricing
COMPARE
30 316L Stainless Steel 45
$646.00USD / Each
COMPARE
60 316L Stainless Steel 110
$772.00USD / Each
COMPARE
100 316L Stainless Steel 190
$982.00USD / Each
COMPARE
150 316L Stainless Steel 300
$1,180.00USD / Each
COMPARE
200 316L Stainless Steel 370
$1,385.00USD / Each
COMPARE
60 Aluminium 80
$229.75USD / Each
COMPARE
100 Aluminium 135
$263.75USD / Each
COMPARE
150 Aluminium 190
$322.50USD / Each
COMPARE
200 Aluminium 245
$360.50USD / Each
COMPARE
250 Aluminium 350
$435.00USD / Each
Showing 1- 10 of 10