Cole-Parmer® Specific Gravity ASTM Hydrometers for Liquids Heavier than Water
  • NIST Traceable
  • Temperature of Standardization 60°F/60°F
  • Conform to ASTM Specifications
  • Metal Ballast
  • Individually Serialized
These hydrometers conform to the accuracy requirements for ASTM hydrometers. Scales are affixed with clear epoxy. Each hydrometer is calibrated in a triple accredited ISO/IEC 17025:2005, A2LA accredited laboratory against equipment whose calibration is traceable to NIST. Individually packaged with a multi-language Statement of Accuracy indicating accuracy traceable to NIST.
ITEM(S) ON SALE
$62.64 - $88.40USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
1.000/1.050 Specific Gravity ASTM 125H Hydrometer for Heavy Liquids
$62.64USD / Each
$73.70USD / Each
COMPARE
1.050/1.100 Specific Gravity ASTM 126H Hydrometer for Heavy Liquids
$64.77USD / Each
$76.20USD / Each
COMPARE
1.150/1.200 Specific Gravity ASTM 128H Hydrometer for Heavy Liquids
$65.79USD / Each
$77.40USD / Each
COMPARE
1.200/1.250 Specific Gravity ASTM 129H Hydrometer for Heavy Liquids
$66.72USD / Each
$78.50USD / Each
COMPARE
1.250/1.300 Specific Gravity ASTM 130H Hydrometer for Heavy Liquids
$70.72USD / Each
$83.20USD / Each
COMPARE
1.550/1.600 Specific Gravity ASTM 136H Hydrometer for Heavy Liquids
$88.40USD / Each
$104.00USD / Each