Cole-Parmer® Specific Gravity & Baume Dual Scale Hydrometers for for Liquids Heavier than Water
  • NIST Traceable
  • Temperature of Standardization 60°F/60°F
  • Metal Ballast
  • Individually Serialized
For liquids heavier than water, each hydrometer is calibrated in a triple accredited ISO/IEC 17025:2005, A2LA accredited laboratory against equipment whose calibration is traceable to NIST. Individually packaged with a multi-language Statement of Accuracy indicating accuracy traceable to NIST.
ITEM(S) ON SALE
$30.77 - $43.52USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Division Hydrometer Form Specific Gravity Range (Max) (%)
Availability Pricing
COMPARE
0.020 SG / 0.2° Baume Plain 2
$30.77USD / Each
$36.20USD / Each
COMPARE
0.005 SG / 0.5° Baume Plain 1.225
$38.25USD / Each
$45.00USD / Each
COMPARE
0.005 SG / 0.5° Baume Plain 1.425
$43.52USD / Each
$51.20USD / Each
COMPARE
0.010 SG / 1.0° Baume Plain 2
$39.69USD / Each
$46.70USD / Each