Cole-Parmer® Spectroscopy Standards Kits for UV-Visible Spectrophotometers
Easily and accurately perform certified spectrophotometer evaluations in any visible or UV-visible spectrophotometer!
  • Standards are durable, scratch-resistant and never need to be replaced
  • ISO 17025 accredited for testing and calibration and NIST compatible
Verify the performance and accuracy of your spectrophotometer right in your own lab. These permanent standards do not have to be recalibrated due to material aging. Unique material construction is scratch resistant and provides stable optical performance.
$3,280.00 - $9,730.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Photometric Accuracy Stray Light Description
Availability Pricing
COMPARE
Yes Yes UV/Vis Photometric Accuracy and Stray Light Calibration Kit
$5,745.00USD / Each
COMPARE
Yes No Photometric Accuracy Calibration Kit for Spectroscopy
$3,280.00USD / Each
COMPARE
Yes Yes Photometric Accuracy and Stray Light Calibration Wavelength Kit
$9,730.00USD / Each