Cole-Parmer® Standard Series Flammable Material Storage Freezers
Safe and reliable freezer storage for flammable materials
  • Meets NFPA 45 and 70 guidelines designed for safety with sealed electrical components
  • Digital microprocessor ensures accurate temparature settings from -15°C to -25°C for critical materials
  • Secure and durable design with a keyed door lock, probe access port, and a magnetic door gasket for a tight seal
  • Uses natural hydrocarbon refrigerants for a sustainable choice
Cole-Parmer® Standard Series Flammable Material Storage Freezers provide a purpose-built solution for laboratories requiring safe, compliant storage of flammable materials. Designed to meet NFPA and OSHA safety guidelines, these units feature a sealed, vapor-proof compressor enclosure and no interior electrical components, minimizing ignition risks. A digital microprocessor temperature controller ensures precise management of storage conditions, along with audible and visual alarms that alert users when temperature is out of a set range. With security features like a keyed door lock and magnetic gasket, plus a durable design backed by a two-year warranty, these freezers offer both safety and reliability for general laboratory use.
ITEM(S) ON SALE
$2,292.00 - $4,824.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Freezer Capacity (cu ft) Min Temperature - Freezer (° C) Power (VAC)
Availability Pricing
COMPARE
14 -25 120
$3,816.00USD / Each
$4,770.00USD / Each
COMPARE
17 -25 120
$4,252.00USD / Each
$5,315.00USD / Each
COMPARE
20 -25 120
$4,824.00USD / Each
$6,030.00USD / Each
COMPARE
4 -25 120
$2,292.00USD / Each
$2,865.00USD / Each