Cole-Parmer® Standard Series Flammable Material Storage Refrigerators
Safe and reliable cold storage for flammable materials
  • Meets NFPA 45 and 70 guidelines designed for safety with sealed electrical components
  • Digital microprocessor ensures accurate temparature settings from 1°C to 10°C for critical materials
  • Secure and durable design with a keyed door lock, probe access port, and a magnetic door gasket for a tight seal
  • Uses natural hydrocarbon refrigerants for a sustainable choice
Cole-Parmer® Standard Series Flammable Material Storage Refrigerators provide a purpose-built solution for laboratories requiring safe, compliant storage of flammable materials. Designed to meet NFPA and OSHA safety guidelines, these units feature a sealed, vapor-proof compressor enclosure and no interior electrical components, minimizing ignition risks. A digital microprocessor temperature controller ensures precise management of storage conditions, along with audible and visual alarms that alert users when temperature is out of a set range. With security features like a keyed door lock and magnetic gasket, plus a durable design backed by a two-year warranty, these refrigerators offer both safety and reliability for general laboratory use.
ITEM(S) ON SALE
$2,400.00 - $4,352.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Min Temperature - Refrigerator (° C) Power (VAC) Refrigerator Capacity (cu ft)
Availability Pricing
COMPARE
1 120 4
$2,400.00USD / Each
$3,000.00USD / Each
COMPARE
1 120 14
$3,776.00USD / Each
$4,720.00USD / Each
COMPARE
1 120 17
$4,020.00USD / Each
$5,025.00USD / Each
COMPARE
1 120 20
$4,352.00USD / Each
$5,440.00USD / Each