Cole-Parmer® Standard Series Hazardous Location Freezers
Reliable cold storage for hazardous environments
  • Microprocessor temperature controller maintains stable conditions from -15°C to -25°C
  • Built for safety - Meets NFPA and OSHA guidelines for Hazardous Locations, Class 1, Division II, Group C & D
  • Advanced monitoring and security with digital display, audible/visual alarms, and remote alarm contacts for enhanced product protection
  • Environmentally friendly refrigerants, magnetic door gaskets, and cycle defrost for long-term reliability
Engineered for hazardous locations, these freezers offer precise temperature management and enhanced security features. A microprocessor temperature controller ensures stable low-temperature storage, while an advanced alarm system provides real-time monitoring for critical samples. Built to meet NFPA and OSHA standards, these units are designed for safe operation in volatile environments. With a rugged, hard-wired construction and eco-friendly refrigerants, they deliver reliable performance in demanding settings.
ITEM(S) ON SALE
$4,804.00 - $5,224.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Freezer Capacity (cu ft) Max Temperature - Freezer (° F) Min Temperature - Freezer (° F)
Availability Pricing
COMPARE
14 5 -13
$4,824.00USD / Each
$6,030.00USD / Each
COMPARE
17 5 -13
$4,804.00USD / Each
$6,005.00USD / Each
COMPARE
20 5 -13
$5,224.00USD / Each
$6,530.00USD / Each