Cole-Parmer® Sterile Serological Pipette and Organizing Rack Bundle
Protect pipettes and minimize clutter
  • Bundle combines serological pipettes and organizing rack for added convenience
  • Pipettes are sterilized by gamma irradiation and individually wrapped to eliminate cross contamination
  • Safely store and protect your pipettes with this durable acrylic rack
These Cole-Parmer® serological pipettes and organizing rack are bundled for exceptional convenience. Heavy-duty, blue-tinted, acrylic rack features five angled compartments which can house 1 to 100 mL glass or plastic pipettes. Two slots at the top of the rack are designed to hold pipette controllers. Organizing rack has exterior dimensions of 3.5” W x 15.875” H x 11.5” D.
These serological pipettes are made from transparent polystyrene (PS) and sterilized by gamma irradiation. Choose from standard serological pipettes with highly legible permanent graduations or aspirating serological pipettes without graduations and filter plugs for vacuum aspirating and free-flowing applications.
$265.00 - $337.50USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Volume (mL) Package Of Tip Style
Availability Pricing
COMPARE
2 1000 Individually wrapped pipettes Self Aspirating
$334.50USD / Each
COMPARE
100 100 Individually wrapped pipettes Standard
$337.50USD / Each
COMPARE
10 400 Individually wrapped pipettes Standard
$265.00USD / Each
COMPARE
25 300 Individually wrapped pipettes Standard
$316.00USD / Each