Cole-Parmer® Stir Bar Kits
Boxed sets include an assortment of useful sizes in a re-usable container
  • Isostatic encapsulation eliminates cracks and porosity
  • FDA and USP Class VI approved PTFE
  • Polished finish reduces pick-up and cross contamination
Magnetic stirring is a widely used method for stirring and mixing in liquid media. The process is not only simple and inexpensive, but extremely diverse in the range of application.
$141.75 - $201.25USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
PTFE Stirrer Bars, Boxed Assortments, 12 Octahedral, 2 each of 13 x 8, 15 x 8, 25 x 10, 38 x 10, 51 x 10, 64 x 10 mm
$141.75USD / Each
COMPARE
PTFE Stirrer Bars, Boxed Assortments, 12 Oval, 2 each of 10 x 5, 15 x 6, 25 x 10, 30 x 10, 35 x 13, 1 each of 50 x 17, 70 x 27 mm
$190.50USD / Each
COMPARE
PTFE Stirrer Bars, Boxed Assortments, 18 Plain, 2 each of 10 x 6, 15 x 6, 20 x 7, 25 x 8, 30 x 7, 40 x 8, 50 x 8, 60 x 7, 80 x 10 mm
$201.25USD / Each