Cole-Parmer® Transportable Dewar Flasks
Fasten lid with clips to use as a transportable vessel
  • Robust design
  • Dewar flask meets DIN EN ISO 16496
These silvered and evacuated Dewars feature a rugged outer casing made of galvanized steel plate with a baked enamel finish. An insulated synthetic lid seals the vessel and locks for transport with two spring clips. A detachable bail-type aluminium handle makes transportation easy. The Dewar flask is made of borosilicate glass 3.3 ISO 3585. Ideal for cooling small samples.
Note: For dry ice and liquid nitrogen storage, drill a 2-mm hole in the lid.
$532.00 - $819.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Inside Diameter (ID) (cm) Outside Diameter (OD) (cm) Volume (Liters)
Availability Pricing
COMPARE
10 15.9 1
$656.00USD / Each
COMPARE
13.8 16 2
$532.00USD / Each
COMPARE
13.8 16 4
$819.00USD / Each