Cole-Parmer® Upright Freezer Racks for 96-Well & 384-Well Microtiter Plates
Conveniently store microtiter plates
  • Included locking rods keep plates in place
  • Handles on either side for quick storage and retrival of rack
  • Corrosion-resistant stainless steel construction for durability
Racks feature handles at both ends to accommodate freezers with right- or left-handed doors.
$148.50 - $353.50USD / Each
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity Height (in) Length (in)
Availability Pricing
COMPARE
70 plates w/lids or 80 plates w/out lids 9 3/16 18 3/8
$191.75USD / Each
COMPARE
84 plates w/lids or 96 plates w/out lids 9 3/16 22
$234.00USD / Each
COMPARE
42 plates w/lids or 48 plates w/out lids 9 3/16 11
$148.50USD / Each
COMPARE
54 plates w/lids or 60 plates w/out lids 12 11
$167.00USD / Each
COMPARE
56 plates w/lids or 64 plates w/out lids 9 3/16 14 11/16
$161.75USD / Each
COMPARE
64 plates w/lids or 80 plates w/out lids 11 14 11/16
$184.50USD / Each
COMPARE
72 plates w/lids or 84 plates w/out lids 12 14 11/16
$212.25USD / Each
COMPARE
96 plates w/lids or 112 plates w/out lids 16 14 11/16
$249.50USD / Each
COMPARE
85 plates w/lids or 95 plates w/out lids 11 18 3/8
$219.50USD / Each
COMPARE
90 plates w/lids or 100 plates w/out lids 12 18 3/8
$252.50USD / Each
COMPARE
96 plates w/lids or 120 plates w/out lids 11 22
$253.50USD / Each
COMPARE
108 plates w/lids or 132 plates w/out lids 12 22
$296.75USD / Each
COMPARE
144 plates w/lids or 168 plates w/out lids 16 22
$353.50USD / Each
COMPARE
98 plates w/lids or 112 plates w/out lids 9 3/16 25 11/16
$261.75USD / Each
COMPARE
112 plates w/lids or 140 plates w/out lids 11 25 11/16
$292.75USD / Each
COMPARE
126 plates w/lids or 154 plates w/out lids 12 25 11/16
$341.00USD / Each
Showing 1- 16 of 16