Cole-Parmer® Upright Freezer Racks for 96-Well Microtube Boxes or Boxes with 2 1/8" Max Height
Included locking rods securely hold boxes in place
  • Access boxes from either side of rack
  • Handles on both sides accommodate freezers with left- or right-handed door
  • Sturdy corrosion-resistant stainless steel construction for long-term use
Racks feature handles at both ends to accommodate freezers with right- or left-handed doors.
$153.50 - $381.50USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity Height (in) Length (in)
Availability Pricing
COMPARE
3 x 4 Array 8 13/16 11 1/16
$153.50USD / Each
COMPARE
5 x 4 Array 8 13/16 18 3/8
$240.00USD / Each
COMPARE
6 x 4 Array 8 13/16 22
$283.25USD / Each
COMPARE
6 x 5 Array 11 22
$298.75USD / Each
COMPARE
6 x 6 Array 13 3/16 22
$292.75USD / Each
COMPARE
7 x 4 Array 8 13/16 25 11/16
$381.50USD / Each