Cole-Parmer® Vertical/Chest Racks with Spring Clips for Mini 2" Boxes
Spring clips securely hold boxes in rack yet allow for quick and easy removal of boxes
  • Accommodates mini 2" cardboard or plastic boxes
  • Sturdy corrosion-resistant stainless steel construction
  • Easily retrieve rack from freezer or LN tank using sturdy dual handles
Use these vertical racks with a wide variety of chest freezers and liquid nitrogen tanks. Included locking rod provides added security that the boxes will stay in place.
$124.75 - $135.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity Height (in)
Availability Pricing
COMPARE
11 boxes 24 3/16
$124.75USD / Each
COMPARE
12 boxes 26 3/8
$135.00USD / Each
COMPARE
13 boxes 28 1/4
$132.00USD / Each