Cole-Parmer® Vial Storage Liquid Nitrogen Dewars
Perfect for storing cryogenic vials in box-style rack system in liquid or vapor-stage
  • Features rack indexed ring, internal locator, and computer-compatible box storage for simple inventory management
  • Temperature maintained at -190°C at the top of racks in vapor-phase storage
  • Holds up to six racks with 5 shelves each
  • Ribbed aluminum body and magneformed necktube offer durability and structural strength
  • Lockable lid
Store cryogenic vials in liquid or vapor-phase nitrogen in these vial storage dewars. Dewars accommodate six racks with five shelves per rack to store up to 6000 2 mL vials. Simple inventory management features include rack index locating ring, internal location fixture, and computer-compatible box storage.
Super insulated dewars provide superior vacuum performance and maximum holding times. Vapor-phase temperatures maintained as low as -190°C at the top of the racks ensure sample stability. Rugged, high-strength aluminum body and magneformed necktube offer durability. Order vial storage dewars with CS100 controller to monitor the temperature and monitor/control the liquid nitrogen level in your dewar.
$3,060.00 - $10,565.00USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Static Holding Time (days) Volume (Liters)
Availability Pricing
COMPARE
130 35
$3,060.00USD / Each
COMPARE
106 81
$5,845.00USD / Each
COMPARE
130 130
$6,615.00USD / Each
COMPARE
130 130
$9,715.00USD / Each
COMPARE
165 165
$5,860.00USD / Each
COMPARE
165 165
$10,565.00USD / Each