Corning® Gosselin™ Sterile Sampling Dippers with Handle
Removable handle makes sample collection quick and easy
  • Obtain excellent chemical resistance from all polypropylene (PP) construction
The Corning® Gosselin™ dippers are designed with a removable handle, making sample collection even easier. Simply take your sample, remove the handle, and send the shockproof container to your testing facility. Dippers are sterile and individually packed inside a traceable and tamper-evident zipped bag to avoid cross-contamination. Choose from bottles with a screw or hinge cap, ideal for any water sampling application.
$312.00 - $370.00USD / Case of 150
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Cap Configuration Height (cm) Sample Volume
Availability Pricing
Corning Gosselin Dipper with Removable Handle, Polypropylene, Sterile, Hinged Cap, 90 mL; 150/Cs EW-34551-53 Mfr # TAFL1-01
COMPARE
Hinged 27.5 90
$370.00USD / Case of 150
Corning Gosselin Dipper with Removable Handle, Polypropylene, Sterile, Screw Cap, 125 mL; 100/CS EW-34551-76 Mfr # TAT52C-01
COMPARE
Screw 27.5 125
$312.00USD / Case of 100
Corning Gosselin Dipper with Removable Handle, Polypropylene, Sterile, Screw Cap, 180 mL; 100/Cs EW-34551-91 Mfr # TAT53C-01
COMPARE
Screw 30.5 180
$317.00USD / Case of 100