Corning® Pyrex® 51395 UV-Blocking Low Actinic Media Bottles
Low actinic borosilicate
  • Store and transport light-sensitive
  • solutions in the range of 3000 to 5000 Å
  • Will not drip while pouring
  • Autoclavable bottle and PP lid
Made from low actinic, code no. 7740 borosilicate glass with red stain, these heavy-duty bottles are ideal for storing/transporting chemicals sensitive to light in the 3000 to 5000 Å range (300 to 500 nm).
Bottles include a linerless, one-piece autoclavable, orange polypropylene, plug-seal screw cap with drip-free pouring ring. Glass bead indicates full capacity line. Bottles feature white enamel graduations and marking spots. Replacement caps and pouring rings are available (sold separately).
$139.00 - $288.00USD / Case of 4
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Material
Availability Pricing
Corning Pyrex 51395 UV-Blocking Low Actinic Media Bottle, 100 mL; 4/CS
Buy More Save More
EW-34574-21 Mfr # 51395-100
COMPARE
100 20 Borosilicate Glass
$139.00USD / Case of 4
Corning Pyrex 51395 UV-Blocking Low Actinic Media Bottle, 250 mL; 4/CS
Buy More Save More
EW-34574-22 Mfr # 51395-250
COMPARE
250 50 Borosilicate Glass
$240.00USD / Case of 4
Corning Pyrex 51395 UV-Blocking Low Actinic Media Bottle, 500 mL; 4/CS
Buy More Save More
EW-34574-23 Mfr # 51395-500
COMPARE
500 100 Borosilicate Glass
$288.00USD / Case of 4
Corning Pyrex 51395 UV-Blocking Low Actinic Media Bottle, 1 L; 4/CS
Buy More Save More
EW-34574-24 Mfr # 51395-1L
COMPARE
1000 100 Borosilicate Glass
$288.00USD / Case of 4