CRS Large-Capacity Moisture Traps
Quick and easy install and removal without disconnecting fittings from the line
  • Suitable for drying laboratory air or central air supplies
Traps remove moisture, oils, and dust from carrier gases. Large 400-mL capacity means fewer refillings are required. Adsorbent materials are indicating desiccant and 5-Å molecular sieve. Desiccant changes color from blue to pink when depleted.
The silicone O-ring seals maintain pressures to 100 psi. Traps feature transparent acrylic housing. Measure 17"L x 2" dia. Choose a trap with base plate for benchtop mounting. Shpg wt 3 lb (1.4 kg).
$240.00 - $250.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Trajan CRS Large-Capacity Moisture Trap without Base Plate, 400 mL Volume, 1/4" Brass Fittings EW-34651-10 Mfr # 202239XL-B
COMPARE
Large-capacity moisture traps; 400 mL volume; 1/4" fitting; no base plate
$250.00USD / Each
Trajan CRS Large-Capacity Moisture Trap without Base Plate, 400 mL Volume, 1/8" Brass Fittings EW-34651-20 Mfr # 202238XL-B
COMPARE
Large-capacity moisture traps; 400 mL volume; 1/8" fitting; no base plate
$240.00USD / Each