Cylindrical Metal Dewar Flasks with Handles
This flask incorporates the convenience of a coated handle
  • Increased portability and pouring efficiency
  • Dewar lip is extended to facilitate pouring
  • Vented polyethylene lid helps to minimize evaporation
These Dewars feature an exterior metal housing with a boroscilicate glass interior. There are no pads between walls to eliminate heat transfer potential. These are ideal for temporary storage or for transfer applications. Dewar flasks come with lids.
$304.00 - $1,025.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Volume (Liters)
Availability Pricing
COMPARE
1.9
$935.00USD / Each
COMPARE
4.3
$1,025.00USD / Each
COMPARE
1
$304.00USD / Each