Davis Instruments Weather Envoy
Download current reading from your Davis Weather Stations for PC analysis.
Use Weather Envoy and WeatherLink software to easily download current weather readings from your Davis Vantage Pro2 or Vantage Vue weather station to your PC or Mac. It can also retransmit data to the console, effectively extending your station’s range. Weather Envoy includes its own barometer and inside temperature and humidity sensors. This product comes in wireless and cabled versions, and both include an AC-power adapter. WeatherLink software is required and sold separately.
$309.75 - $404.25USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Connections Compatible With
Availability Pricing
Davis Instruments Wireless Weather Envoy EW-30003-60 Mfr # 6316
COMPARE
Wireless Vantage Pro2 or Vantage Vue weather station
$309.75USD / Each
Davis Instruments Cabled Weather Datalogger EW-30003-61 Mfr # 6316C
COMPARE
Cabled Cabled Vantage Pro2 or Vantage Pro2 Plus weather station sensor suite
$404.25USD / Each