Davis Instruments Wireless Leaf-Soil Moisture Temperature Station
Remotely monitor leaf, and soil moisture and temperature
  • Operates under solar power
The Wireless Leaf & Soil Moisture/Temperature Station for Vantage Pro2™ monitors up to four temperature compensated Soil Moisture Sensors and two Leaf Wetness Sensors anywhere within the transmission range of your wireless Vantage Pro2 console. The station communicates directly to your Wireless Vantage Pro2 console/receiver over any one of eight user-selectable ID codes, and has a transmitting range of between 200' to 1000' (75 to 300 m) depending upon the environment. The station is solar powered, and includes a battery backup.
2 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (Outside) (° F) Temperature Accuracy (Outside) (° F) Temperature Resolution (Outside) (° F)
Availability Pricing
Davis Instruments 6345 Leaf/Soil Moisture/Temp Monitoring Station; w/o Sensors EW-32045-03 Mfr # 6345
COMPARE
150
$841.05USD / Each
Davis Instruments 6345CS Leaf/Soil Moisture/Temp Monitoring Station; w/ Sensors EW-32245-49 Mfr # 6345CS
COMPARE
150
$1,051.05USD / Each